Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GMPR Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548473

Catalog No. PA548473

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes an enzyme that catalyzes the irreversible and NADPH-dependent reductive deamination of GMP to IMP. The protein also functions in the re-utilization of free intracellular bases and purine nucleosides.
Specifications

Specifications

GMPR
Polyclonal
PBS with 0.09% sodium azide
P36959
GMP reductase 1, Guanosine 5′-monophosphate oxidoreductase 1, Guanosine monophosphate reductase 1, GMPR, GMPR1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2766
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
GMPR
Liquid
GMPR
KLH conjugated synthetic peptide between 238-267 amino acids from the C-terminal region of human GMPR.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit