Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GMPR2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP15283720UL

 View more versions of this product

Catalog No. NBP15283720


Add to cart

Description

Description

GMPR2 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

GMPR2
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Purified
GMPR2
Synthetic peptides corresponding to GMPR2(guanosine monophosphate reductase 2) The peptide sequence was selected from the C terminal of GMPR2. Peptide sequence GAGADFVMLGGMLAGHSESGGELIERDGKKYKLFYGMSSEMAMKKYAGGV.
Protein A purified
RUO
Primary
51292
Western Blot
Western Blot 1:100-1:2000
Q9P2T1
EC 1.7.1.7, GMP reductase 2, Guanosine 5'-monophosphate oxidoreductase 2, guanosine monophosphate reductase 2MGC830, guanosine monophosphate reductase isolog, MGC15084
Rabbit
IgG
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit