Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GMPS Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549552

Catalog No. PA549552

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Involved in the de novo synthesis of guanine nucleotides which are not only essential for DNA and RNA synthesis, but also provide GTP, which is involved in a number of cellular processes important for cell division.
Specifications

Specifications

GMPS
Polyclonal
PBS with 0.09% sodium azide
P49915
GMP synthase [glutamine-hydrolyzing], GMP synthetase, Glutamine amidotransferase, GMPS
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
8833
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
GMPS
Liquid
GMPS
KLH conjugated synthetic peptide between 602-630 amino acids from the C-terminal region of human GMPS.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit