Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris™ GMQ hydrochloride

ASIC3 channel opener

Manufacturer:  Tocris™ 5028/10

 View more versions of this product

Catalog No. 502810R


  Check Availability
Add to cart

Disclaimers:

For Research Use Only


Specifications

Description

Selective opener of acid-sensing ion channel 3 (ASIC3) at pH 7.4. Alters the pH dependence of ASIC1a and ASIC1b; does not affect the activation curve of ASIC2a.
  • Chemical Name: N-(4-Methyl-2-quinazolinyl)-guanidine hydrochloride
  • Formula: C10H11N5.HCl
  • CAS Number: 5361-15-9
  • Solubility: Soluble to 50mM in DMSO and to 5mM in water with gentle warming
  • Purity: >98
Description & Specifications

Specifications

GMQ hydrochloride
>98
10mg
GMQ hydrochloride
237.69
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit