Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GN 44028 Available on GSA/VA Contract for Federal Government customers only.

Potent HIF-1α inhibitor

$205.00 - $844.50

Specifications

Chemical Name or Material N-(2,3-Dihydro-1,4-benzodioxin-6-yl)-1,4-dihydroindeno[1,2-c]pyrazol-3-amine
Molecular Formula C18N15N3O2
CAS 1421448-26-1
Product Type Target: HIF Inhibitors
Formula Weight 305.33
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
56-551-0 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5655/10
10mg Each for $205.00
Add to cart
 
56-555-0 View Documents Tocris Bioscience™
5655/50
50mg Each for $844.50
Add to cart
 
Description

Description

Potent HIF-1α inhibitor (IC50 = 14 nM). Inhibits hypoxia-induced HIF-1α transcriptional activity without suppressing HIF-1α mRNA expression, HIF-1α protein accumulation, or HIF-1α/HIF-1β heterodimerization. Cytotoxic to a range of cancer cell types in vitro.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

N-(2,3-Dihydro-1,4-benzodioxin-6-yl)-1,4-dihydroindeno[1,2-c]pyrazol-3-amine
1421448-26-1
305.33
C18N15N3O2
Target: HIF Inhibitors
Safety and Handling

Safety and Handling

Recommended Storage : Store at 4°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit