Learn More
Aviva Systems Biology Corp GNA15 Antibody - N-terminal region (ARP58472_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP58472_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GNA15. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GNA15. Alternate names: GNA16ApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-62318