Learn More
GNA15 Mouse anti-Human, Clone: OTI1D3, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF808159
Catalog No. 50-166-8660
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Guanine nucleotide-binding proteins (G proteins) are involved as modulators or transducers in various transmembrane signaling systems.
Specifications
Specifications
GNA15
Monoclonal
Unconjugated
GNA15
Lyophilized
GNA15
Human recombit protein fragment corresponding to amino acids 1-256 of human GNA15 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI1D3
PBS with 8% trehalose and no preservative; pH 7.3
P30679
GNA16
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
2769