Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GNAQ Mouse anti-Human, Clone: GNAQ/2434, Novus Biologicals™
Mouse Monoclonal Antibody
$265.00
Specifications
Antigen | GNAQ |
---|---|
Clone | GNAQ/2434 |
Concentration | 0.2 mg/ml |
Dilution | Flow Cytometry 1-2 ug/million cells, ELISA, Immunocytochemistry/Immunofluorescence 1-2 ug/ml |
Applications | Flow Cytometry, ELISA, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB122100
|
Novus Biologicals
NBP30731320UG |
20 μg |
Each of 1 for $265.00
|
|
|||||
Description
GNAQ Monoclonal specifically detects GNAQ in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
GNAQ | |
0.2 mg/ml | |
Flow Cytometry, ELISA, Immunofluorescence | |
Unconjugated | |
Mouse | |
Human | |
GAQG-ALPHA-q, guanine nucleotide binding protein (G protein), q polypeptide, Guanine nucleotide-binding protein alpha-q, guanine nucleotide-binding protein G(q) subunit alpha | |
Recombinant full-length human GNAQ protein | |
Primary | |
Store at 4°C. |
GNAQ/2434 | |
Flow Cytometry 1-2 ug/million cells, ELISA, Immunocytochemistry/Immunofluorescence 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
2776 | |
IgG2b κ | |
Protein A or G purified |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title