Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GNAT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550982

Catalog No. PA550982


Add to cart

Description

Description

GNAT1 Polyclonal Antibody detects endogenous levels of GNAT1 protein.

Transducin is a 3-subunit guanine nucleotide-binding protein (G protein) which stimulates the coupling of rhodopsin and cGMP-phoshodiesterase during visual impulses. The transducin alpha subunits in rods and cones are encoded by separate genes. This gene encodes the alpha subunit in rods. This gene is also expressed in other cells, and has been implicated in bitter taste transduction in rat taste cells. Mutations in this gene result in autosomal domit congenital stationary night blindness. Multiple alternatively spliced variants, encoding the same protein, have been identified.
Specifications

Specifications

GNAT1
Polyclonal
Unconjugated
GNAT1
Liquid
GNAT1
Synthesized peptide derived from the Internal region of human G&No.945; t1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
ELISA, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
P11488, P20612
GNAT1, GNATR, Guanine nucleotide-binding protein G(t) subunit alpha-1, Transducin alpha-1 chain
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
14685, 2779, 363143
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit