Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GNAT2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA524553
Description
GNAT2 Polyclonal Antibody for Western Blot, IHC (P)
Transducin is a 3-subunit guanine nucleotide-binding protein (G protein) which stimulates the coupling of rhodopsin and cGMP-phoshodiesterase during visual impulses. The transducin alpha subunits in rods and cones are encoded by separate genes. This gene encodes the alpha subunit in cones.
Specifications
GNAT2 | |
Polyclonal | |
Unconjugated | |
Gnat2 | |
ACHM4; AW490837; cone-type transducin alpha subunit; G protein subunit alpha transducin 2; Gnat2; Gnat-2; GNATC; Gt-2; guanine nucleotide binding protein (G protein), alpha transducing 2; guanine nucleotide binding protein (G protein), alpha transducing activity polypeptide 2; guanine nucleotide binding protein, alpha transducing 2; guanine nucleotide-binding protein G(t) subunit alpha-2; RP5-1160K1.4; Tcalpha; Transducin alpha-2 chain; transducin, cone-specific, alpha polypeptide | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
14686, 2780 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P19087, P50149 | |
Gnat2 | |
KLH conjugated synthetic peptide between 140-169 amino acids from the central region of human GNAT2. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction