Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GNB2L1 Mouse anti-Human, Clone: OTI2D8, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA502717

Catalog No. 50-167-3749


Add to cart

Description

Description

RACK1 (receptor for activated C kinase 1) was identified through its binding to various PKC isoforms. Its main function is to recruit PKC and various other proteins to specific locations to form multiprotein complexes, mediating various signal pathways. RACK1 is required for VANGL2 membrane localization and for PTK2/FAK1 phosphorylation and dephosphorylation.
Specifications

Specifications

GNB2L1
Monoclonal
0.39 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P63244
GNB2L1, Gnb2-rs1, H12.3, HLC-7, PIG21
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10399
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI2D8
Unconjugated
RACK1
Liquid
RACK1
Full length human recombit protein of human GNB2L1 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit