Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GNB5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109781
Description
Immunogen sequence: MATEGLHENE TLASLKSEAE SLKGKLEEER AKLHDVELHQ VAERVEALGQ FVMKTRRTLK GHGNKVLCMD WCKDKRRIVS SSQDGKVIVW DSFTTNKEHA VTMPCTWVMA CAYAPSGCAI ACGGLDNKCS VYPLTFDKNE NMAAKKKSVA MHTNYLSACS FTNSDMQILT ASGDGTCALW DVESGQLLQS FHGHGADVLC.
The Gb5 protein is a G protein subunit present predominantly in the central nervous system. This protein appears to be expressed in two forms, one found in the brain of 39 kDa and one found in the retina of 44 kDa. It has also been found that Gb5 associates tightly with RGS7, a member of the superfamily of negative regulators of G protein signaling. These complexes have been localized to most regions of the brain, whereas Gb1-4 exhibit no observable colocalization with RGS7, despite their widespread codistribution throughout similar neuronal networks. This antibody is specific for a peptide sequence within the D1 blade which is not found in the Gb's 1-4.
Specifications
GNB5 | |
Polyclonal | |
Unconjugated | |
Gnb5 | |
flail; flailer; flr; G protein subunit beta 5; G protein, beta subunit 5L; G protein, beta-5 subunit; GB5; GBB5; Gbeta5; GBS; Gnb5; guanine nucleotide binding protein (G protein), beta 5; guanine nucleotide binding protein (G protein), beta polypeptide 5; guanine nucleotide binding protein, beta 5; guanine nucleotide-binding protein subunit beta-5; guanine nucleotide-binding protein, beta subunit 5L; guanine nucleotide-binding protein, beta-5 subunit; hug; transducin beta chain 5 | |
Rabbit | |
Affinity chromatography | |
RUO | |
10681, 14697, 83579 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.65 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O14775, P62881, P62882 | |
Gnb5 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human GNB5 (NP_006569.1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction