Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GNB5 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP258164

Catalog No. NBP258164


Add to cart

Description

Description

GNB5 Polyclonal antibody specifically detects GNB5 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

GNB5
Unconjugated
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
FLJ37457, FLJ43714, G protein, beta subunit 5L, G protein, beta-5 subunit, GB5, gbeta5, guanine nucleotide binding protein (G protein), beta 5, guanine nucleotide-binding protein subunit beta-5, guanine nucleotide-binding protein, beta subunit 5L, Transducin beta chain 5
Rabbit
IgG
100 ul
Signal Transduction
Primary
10681
Human
Immunocytochemistry, Immunofluorescence
Immunocytochemistry/Immunofluorescence 1-4 ug/ml
Affinity Purified
GNB5
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:DNKCSVYPLTFDKNENMAAKKKSVAMHTNYLSACSFTNSDMQILTASGDGT
Affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only