Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gng12 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549530

Catalog No. PA549530


Add to cart

Description

Description

Predicted to react with bovine and mouse based on sequence homology.

Guanine nucleotide-binding proteins (G proteins) are involved as a modulator or transducer in various transmembrane signaling systems. The beta and gamma chains are required for the GTPase activity, for replacement of GDP by GTP, and for G protein- effector interaction.
Specifications

Specifications

GNG12
Polyclonal
Unconjugated
GNG12
Liquid
GNG12
KLH conjugated synthetic peptide between 37-68 amino acids from human GNG12.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q9UBI6, Q9DAS9
Guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-12, GNG12
Rabbit
IgG
200 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
14701, 55970
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit