Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GNG13 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104430
Description
Antibody detects endogenous levels of total GNG13.
In mammals, G protein α, β and γ polypeptides are encoded by at least 16, 4 and 7 genes, respectively. Most interest in G proteins has been focused on their α subunits, since these proteins bind and hydrolyze GTP and most obviously regulate the activity of the best studied effectors. Evidence, however, has established an important regulatory role for the βγ subunits. It is becoming increasingly clear that different G protein complexes expressed in different tissues carry structurally distinct members of the γ as well as the α and β subunits, and that preferential associations between members of subunit families increase G protein functional diversity.
Specifications
GNG13 | |
Polyclonal | |
Unconjugated | |
GNG13 | |
1500031D04Rik; AB030194; G gamma subunit, clone 2-35; G gamma subunit, clone:h2-35; G protein subunit gamma 13; G(gamma)13; Ggamma13; Gng13; guanine nucleotide binding protein (G protein), gamma 13; guanine nucleotide binding protein 13, gamma; guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-13; h2-35 | |
Rabbit | |
Affinity chromatography | |
RUO | |
51764, 64337, 685451 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9JMF3, Q9P2W3 | |
GNG13 | |
A synthesized peptide derived from human GNG13(Accession Q9P2W3), corresponding to amino acid residues M1-L35. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction