Learn More
GNG4 Mouse anti-Human, Clone: 1D9-G11, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA524407
Catalog No. PIMA524407
Add to cart
Description
Description
Peptide Sequence: MKEGMSNNST ASISQARKAV EQLKMEACMD RVKVSQAAAD LLAYCEAHVR EDPLIIPVPA SENPFREKKF FCTIL
Guanine nucleotide-binding proteins (G proteins) are involved as a modulator or transducer in various transmembrane signaling systems. The beta and gamma chains are required for the GTPase activity, for replacement of GDP by GTP, and for G protein-effector interaction.
Specifications
Specifications
GNG4
Monoclonal
Unconjugated
GNG4
Liquid
GNG4
GNG4 (AAH22485, 1 a.a. ∽ 75 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Protein A
RUO
Antibody
Monoclonal
Human
ELISA, Western Blot
1D9-G11
PBS with no preservative; pH 7.4
P50150
GNG4; GNGT4
Mouse
IgG1, kappa
100 μg
-20° C
Primary
2786