Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GNL2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548771

Catalog No. PA548771

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

GTPase that associates with pre-60S ribosomal subunits in the nucleolus and is required for their nuclear export and maturation.
Specifications

Specifications

GNL2
Polyclonal
PBS with 0.09% sodium azide
Q13823
Nucleolar GTP-binding protein 2, Autoantigen NGP-1, GNL2, NGP1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
29889
Western Blot
Unconjugated
GNL2
Liquid
GNL2
KLH conjugated synthetic peptide between 18-45 amino acids from the N-terminal region of human GNL2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit