Learn More
Aviva Systems Biology Corp GNL3 Antibody - middle region (ARP58827_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP58827_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GNL3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GNL3. Alternate names: C77032, E2IG3, MGC800, NS, NNP47ApplicationWB. Species reactivity: Human, Rat
Catalog No. 50-014-62439