Learn More
GNMT Mouse anti-Human, Clone: OTI8A3, liquid, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner TA501276
Catalog No. 50-167-2422
Add to cart
Description
Description
The protein encoded by this gene is an enzyme that catalyzes the conversion of S-adenosyl-L-methionine to S-adenosyl-L-homocysteine and sarcosine. The encoded protein is found in the cytoplasm and acts as a homotetramer. Defects in this gene are a cause of GNMT deficiency.
Specifications
Specifications
GNMT
Monoclonal
Unconjugated
GNMT
Liquid
GNMT
Full length human recombit protein of human GNMT produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry, Western Blot
OTI8A3
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q14749
HEL-S-182mP
Mouse
IgG3
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
27232