Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GNMT Mouse anti-Human, Clone: 1041CT18.2.3, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524759

Catalog No. PIMA524759

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The protein encoded by this gene is an enzyme that catalyzes the conversion of S-adenosyl-L-methionine to S-adenosyl-L-homocysteine and sarcosine. The encoded protein is found in the cytoplasm and acts as a homotetramer. Defects in this gene are a cause of GNMT deficiency.
Specifications

Specifications

GNMT
Monoclonal
0.92 mg/mL
PBS with 0.09% sodium azide
Q14749
Glycine N-methyltransferase, 2.1.1.20, GNMT
Mouse
IgG2a, kappa
200 μL
-20°C
Primary
27232
Western Blot
1041CT18.2.3
Unconjugated
GNMT
Liquid
GNMT
Recombit protein between 1-295 amino acids from the human GNMT
Protein G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit