Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GNPDA2 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen GNPDA2
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB095013
View Documents
Novus Biologicals
NBP304487100UL
100 μg Each for $320.71
Add to cart
 
NB095014
View Documents
Novus Biologicals
NBP30448720UL
20 μg Each for $138.37
Add to cart
 
Description

Description

GNPDA2 Polyclonal antibody specifically detects GNPDA2 in Human, Mouse samples. It is validated for Western Blot
Specifications

Specifications

GNPDA2
Unconjugated
Rabbit
Lipid and Metabolism
PBS with 50% glycerol, pH7.3.
132789
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot
Purified
RUO
Human, Mouse
EC 3.5.99.6, GlcN6P deaminase 2, glucosamine-6-phosphate deaminase 24921523I18Rik, glucosamine-6-phosphate isomerase 2, Glucosamine-6-phosphate isomerase SB52, GNP2SB52, GNPDA 2, putative glucosamine-6-phosphate isomerase
Recombinant fusion protein containing a sequence corresponding to amino acids 1-190 of human GNPDA2 (NP_612208.1).
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit