Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GnRH2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552076
Description
Immunogen sequence: HWSHGWYPGG KRALSSAQDP QNALRPPAGS PVQTAHGLPS DALAPLDDSM PWEGRTTAQW SLHRKRHLAR TLLTAAREPR Highest antigen sequence identity to the following orthologs: Mouse - 26%, Rat - 30%.
This gene encodes a secreted peptide hormone and member of the gonadotropin-releasing hormone (GnRH) family of proteins. The encoded protein regulates reproductive function by stimulating the production and release of the gonadotropins follicle-stimulating hormone (FSH) and luteinizing hormone (LH). The encoded protein may inhibit endometrial, ovarian, prostate, and breast cancer cell proliferation. Alternative splicing results in multiple transcript variants.
Specifications
GnRH2 | |
Polyclonal | |
Unconjugated | |
GNRH2 | |
GnRH II; GNRH2; GnRH-associated peptide 2; GnRH-associated peptide II; GnRH-II; Gonadoliberin II; Gonadoliberin-2; gonadotropin releasing hormone 2; gonadotropin-releasing hormone 2; Gonadotropin-releasing hormone II; LH-RH II; LH-RHII; luliberin II; Luteinizing hormone-releasing hormone II; progonadoliberin II; Progonadoliberin-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2797 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O43555 | |
GNRH2 | |
Recombinant protein corresponding to Human GnRH2. Recombinant protein control fragment (Product #RP-101638). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction