Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GnRHR Rabbit anti-Human, Clone: GNRHR/2982R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen GnRHR
Clone GNRHR/2982R
Concentration 0.2 mg/ml
Dilution ELISA
Applications ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122968
View Documents
Novus Biologicals
NBP30774720UG
20 μg
Each for $159.82
Add to cart
 
NB122967
View Documents
Novus Biologicals
NBP30774710UG
100 μg
Each for $406.52
Add to cart
 
Description

Description

GnRHR Monoclonal antibody specifically detects GnRHR in Human samples. It is validated for ELISA
Specifications

Specifications

GnRHR
0.2 mg/ml
ELISA
Purified
RUO
10 mM PBS with 0.05% BSA
2798
IgG
Primary
Store at 4°C.
GNRHR/2982R
ELISA
Unconjugated
Rabbit
Human
GnRH receptor, GnRH-R, GNRHR1, gonadotropin-releasing hormone (type 1) receptor 1, gonadotropin-releasing hormone receptor, GRHR, leutinizing-releasing hormone receptor, LHRHRleutinizing hormone releasing horomone receptor, LRHR, luliberin receptor, type I GnRH receptor
Recombinant full-length human GnRHR protein
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit