Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9(S)-HODE, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89137635 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-635 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-635 Supplier Abnova Corporation Supplier No. PAB13605
Add to Cart
Add to Cart

Goat polyclonal antibody raised against 9(S)-HODE.

Specifications

Antigen 9(S)-HODE
Applications ELISA, Radioimmune Assays (RIA)
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against 9(S)-HODE.
Dilution ELISA (2 ug/well) Radioimmunoassay (2 ug/well) The optimal working dilution should be determined by the end user.
Formulation In PBS
Host Species Goat
Immunogen 9 (S)-Hydroxyoctadecadienoic (HODE) Acid.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Goat antisera specific to 9 (S)-Hydroxyoctadecadienoic (HODE) Acid. HAPTEN REACTIVITY: 9(S)-HODE 100.0% 9(S)-HODE 100.0% 9(R)-HODE 100.0% 13(S)-HODE 1.2% 13(R)-HODE 1.2% 9-oxo-ODE 1.2% 13-oxo-ODE 2.4% 11(S)-HETE 0.0% 15(S)-HETE 0.0% Linoleic Acid 0.0%.
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.