Learn More
ABCD2, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of ABCD2.
Supplier: Abnova Corporation PAB25454
Description
The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the ALD subfamily, which is involved in peroxisomal import of fatty acids and/or fatty acyl-CoAs in the organelle. All known peroxisomal ABC transporters are half transporters which require a partner half transporter molecule to form a functional homodimeric or heterodimeric transporter. The function of this peroxisomal membrane protein is unknown; however this protein is speculated to function as a dimerization partner of ABCD1 and/or other peroxisomal ABC transporters. Mutations in this gene have been observed in patients with adrenoleukodystrophy, a severe demyelinating disease. This gene has been identified as a candidate for a modifier gene, accounting for the extreme variation among adrenoleukodystrophy phenotypes. This gene is also a candidate for a complement group of Zellweger syndrome, a genetically heterogeneous disorder of peroxisomal biogenesis. [provided by RefSeq
Sequence: CPLSDTLAIKGKVIDSpecifications
ABCD2 | |
Polyclonal | |
Unconjugated | |
ELISA (1:32000) Western Blot (0.5-2 ug/mL) The optimal working dilution should be determined by the end user. | |
ABCD2 | |
ABC39/ALDL1/ALDR/ALDRP/hALDR | |
Goat | |
Antigen affinity purification | |
RUO | |
225 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of ABCD2. | |
In Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
NP_005155.1 | |
ABCD2 | |
A synthetic peptide corresponding to amino acids 460-473 at internal region of human ABCD2. | |
100 μg | |
Primary | |
Human | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only