Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACO2 Goat anti-Human, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of ACO2.

Manufacturer:  Abnova Corporation PAB18540

Catalog No. 89-120-099


Add to cart

Description

Description

The protein encoded by this gene belongs to the aconitase/IPM isomerase family. It is an enzyme that catalyzes the interconversion of citrate to isocitrate via cis-aconitate in the second step of the TCA cycle. This protein is encoded in the nucleus and functions in the mitochondrion. It was found to be one of the mitochondrial matrix proteins that are preferentially degraded by the serine protease 15(PRSS15), also known as Lon protease, after oxidative modification. [provided by RefSeq

Sequence: C-QDTYQHPPKDSSGQH
Specifications

Specifications

ACO2
0.5 mg/mL
Goat polyclonal antibody raised against synthetic peptide of ACO2.
In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA)
NP_001089.1
ACONM|MGC20605|MGC33908
Goat
Antigen affinity purification
RUO
Primary
50
ELISA, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
ELISA (1:16000) Western Blot (0.01-0.03 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (3-5 ug/mL) The optimal working dilution should be determined by the end user.
ACO2
Liquid
ACO2
A synthetic peptide corresponding to internal region of human ACO2.
100 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only