Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APOC3, Goat, HRP, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against native APOC3.

Manufacturer:  Abnova Corporation PAB14377

Catalog No. 89-117-023


Add to cart

Description

Description

Apolipoprotein C-III is a very low density lipoprotein (VLDL) protein. APOC3 inhibits lipoprotein lipase and hepatic lipase; it is thought to delay catabolism of triglyceride-rich particles. The APOA1, APOC3 and APOA4 genes are closely linked in both rat and human genomes. The A-I and A-IV genes are transcribed from the same strand, while the A-1 and C-III genes are convergently transcribed. An increase in apoC-III levels induces the development of hypertriglyceridemia. [provided by RefSeq

Specifications

Specifications

APOC3
HRP
ELISA (1:100-1:4000) Western Blot (1:100-1:3000) The optimal working dilution should be determined by the end user.
APOC3
APOCIII|MGC150353
Goat
500 uL
Store at -20°C.
After reconstitution with 500 uL of deionized water and add equal volume of glycerol, store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
This antibody can be used for detection of APOC3 in plasma and Lipoproteins.
ELISA, Western Blot
Goat polyclonal antibody raised against native APOC3.
In 50mM PBS, 100mM NaCl, pH 7.4 (10mg/mL BSA, 0.01% thimerosal)
Lyophilized
APOC3
Native purified human APOC3.
RUO
Primary
345
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only