Learn More
APOE Goat anti-Human, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of APOE.
Supplier: Abnova Corporation PAB6765
Description
Chylomicron remnants and very low density lipoprotein (VLDL) remnants are rapidly removed from the circulation by receptor-mediated endocytosis in the liver. Apolipoprotein E, a main apoprotein of the chylomicron, binds to a specific receptor on liver cells and peripheral cells. ApoE is essential for the normal catabolism of triglyceride-rich lipoprotein constituents. The APOE gene is mapped to chromosome 19 in a cluster with APOC1 and APOC2. Defects in apolipoprotein E result in familial dysbetalipoproteinemia, or type III hyperlipoproteinemia (HLP III), in which increased plasma cholesterol and triglycerides are the consequence of impaired clearance of chylomicron and VLDL remnants. [provided by RefSeq
Sequence: C-VGTSAAPVPSDNHSpecifications
APOE | |
Polyclonal | |
Unconjugated | |
ELISA (1:32000) Western blot (0.1-0.3 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-4 ug/mL) The optimal working dilution should be determined by the end user. | |
APOE | |
AD2/LPG/MGC1571/apoprotein | |
Goat | |
Antigen affinity purification | |
RUO | |
348 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of APOE. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
NP_000032.1 | |
APOE | |
A synthetic peptide corresponding to human APOE. | |
100 μg | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only