Learn More
Avian Influenza Hemagglutinin 3, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of Avian Influenza Hemagglutinin 3.
Manufacturer:  Abnova Corporation PAB12966
Catalog No. 89-137-572
Add to cart
Description
Description
Specifications
Specifications
Avian Influenza Hemagglutinin 3
Unconjugated
ELISA (1 ug/mL) The optimal working dilution should be determined by the end user.
AAT76166
Goat
100 ug
Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
ELISA, Western Blot
Goat polyclonal antibody raised against synthetic peptide of Avian Influenza Hemagglutinin 3.
In PBS (0.02% sodium azide)
Liquid
A synthetic peptide corresponding to internal region 14 amino acids of Avian Influenza Hemagglutinin 3.
RUO
Primary
Influenza virus