Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BDH2 Goat anti-Human, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of BDH2.

Supplier:  Abnova Corporation PAB7523

Catalog No. 89-133-331


Add to Cart

Description

Description

Sequence: C-TKKKQIDQFANE
Specifications

Specifications

BDH2
Polyclonal
Unconjugated
ELISA (1:64000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user.
BDH2
DHRS6/EFA6R/FLJ13261/PRO20933/SDR15C1/UCPA-OR/UNQ6308
Goat
Antigen affinity purification
RUO
56898
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
ELISA, Western Blot
0.5 mg/mL
Goat polyclonal antibody raised against synthetic peptide of BDH2.
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
NP_064524.3
BDH2
A synthetic peptide corresponding to internal region of of human BDH2.
100 μg
Primary
Human
Liquid
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only