Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIRC6 Goat, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of BIRC6.

Manufacturer:  Abnova Corporation PAB19001

Catalog No. 89-120-554


Add to cart

Description

Description

This gene encodes a protein with a BIR (baculoviral inhibition of apoptosis protein repeat) domain and a UBCc (ubiquitin-conjugating enzyme E2, catalytic) domain. This protein inhibits apoptosis by facilitating the degradation of apoptotic proteins by ubiquitination. [provided by RefSeq

Sequence: C-SREYDGNIRQAT
Specifications

Specifications

BIRC6
0.5 mg/mL
Goat polyclonal antibody raised against synthetic peptide of BIRC6.
In 0.5mg/mL in Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
NP_057336.3
APOLLON|BRUCE|FLJ13726|FLJ13786|KIAA1289
Goat
Antigen affinity purification
RUO
Primary
57448
ELISA
Unconjugated
ELISA (1:64000) The optimal working dilution should be determined by the end user.
BIRC6
Liquid
BIRC6
A synthetic peptide corresponding to amino acids at internal region of human BIRC6.
100 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only