Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BPNT1, Goat, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of BPNT1.

Supplier:  Abnova Corporation PAB7352

Catalog No. 89-133-173


Add to Cart

Description

Description

BPNT1, also called bisphosphate 3-prime-nucleotidase, or BPntase, is a member of a magnesium-dependent phosphomonoesterase family. Lithium, a major drug used to treat manic depression, acts as an uncompetitive inhibitor of BPntase. The predicted human protein is 92% identical to mouse BPntase. BPntase's physiologic role in nucleotide metabolism may be regulated by inositol signaling pathways. The inhibition of human BPntase may account for lithium-induced nephrotoxicity. [provided by RefSeq

Sequence: C-RNYDYYASRVPESIK
Specifications

Specifications

BPNT1
Polyclonal
Unconjugated
ELISA (1:4000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user.
BPNT1
PIP
Goat
Antigen affinity purification
RUO
10380
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
ELISA, Western Blot
0.5 mg/mL
Goat polyclonal antibody raised against synthetic peptide of BPNT1.
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
NP_006076.4
BPNT1
A synthetic peptide corresponding to human BPNT1.
100 μg
Primary
Human
Liquid
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only