Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C14orf68 Goat anti-Human, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of C14orf68.

Supplier:  Abnova Corporation PAB7199

Catalog No. 89-133-037


Add to Cart

Description

Description

Sequence: C-KSRLQADGQGQRR
Specifications

Specifications

C14orf68
Polyclonal
Unconjugated
ELISA (1:32000) Immunohistochemistry (2.5 ug/mL) The optimal working dilution should be determined by the end user.
C14orf68
HDMCP/HMFN1655
Goat
Antigen affinity purification
RUO
283600
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
ELISA, Immunohistochemistry (PFA fixed)
0.5 mg/mL
Goat polyclonal antibody raised against synthetic peptide of C14orf68.
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
NP_997000.2
C14orf68
A synthetic peptide corresponding to human C14orf68.
100 μg
Primary
Human
Liquid
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only