Learn More
CACNB4 Goat anti-Human, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of CACNB4.
Supplier: Abnova Corporation PAB6720
Description
This gene encodes a member of the beta subunit family of voltage-dependent calcium channel complex proteins. Calcium channels mediate the influx of calcium ions into the cell upon membrane polarization and consist of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. Various versions of each of these subunits exist, either expressed from similar genes or the result of alternative splicing. The protein encoded by this locus plays an important role in calcium channel function by modulating G protein inhibition, increasing peak calcium current, controlling the alpha-1 subunit membrane targeting and shifting the voltage dependence of activation and inactivation. Certain mutations in this gene have been associated with idiopathic generalized epilepsy (IGE) and juvenile myoclonic epilepsy (JME). Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: CSPGGYSHDSRHRLSpecifications
CACNB4 | |
Polyclonal | |
Unconjugated | |
ELISA (1:8000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user. | |
CACNB4 | |
CAB4/CACNLB4/EA5/EJM | |
Goat | |
Antigen affinity purification | |
RUO | |
785 | |
Human | |
Liquid |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of CACNB4. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
NP_001005747.1;NP_000717.2;NP_001005746.1 | |
CACNB4 | |
A synthetic peptide corresponding to human CACNB4. | |
100 μg | |
Primary | |
This antibody is expected to recognize all reported protein isoforms of human CACNB4 (NP_001005747.1, NP_000717.2, NP_001005746.1, ). | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only