Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Goat anti-Canine IgM Heavy Chain Secondary Antibody, Alkaline Phosphatase, Novus Biologicals™
Goat Polyclonal Antibody
Supplier: Novus Biologicals NB7319
Description
IgM Heavy Chain Polyclonal antibody specifically detects IgM Heavy Chain in Canine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).Specifications
IgM Heavy Chain | |
Polyclonal | |
Alkaline Phosphatase (AP) | |
Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain | |
Dog IgM | |
1 mL | |
Secondary | |
By immunoelectrophoresis and ELISA this reacts specifically with dog IgM. This may cross react with IgM from other species. | |
Store at 4C. Do not freeze. |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
Western Blot 1:5000- 1:50000, ELISA 1:1000 - 1:10000, Immunohistochemistry 1:100 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100 - 1:1000, Immunohistochemistry-Paraffin | |
Goat | |
Affinity Purified | |
RUO | |
3507 | |
Canine | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction