Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CARD17, Goat, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of CARD17.

Manufacturer:  Abnova Corporation PAB17853

Catalog No. 89-119-516


Add to cart

Description

Description

Sequence: C-NHLTTQDSQIVLPS
Specifications

Specifications

CARD17
0.5 mg/mL
Goat polyclonal antibody raised against synthetic peptide of CARD17.
In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA)
NP_001007233.1
INCA
Goat
Antigen affinity purification
RUO
Primary
440068
ELISA, Western Blot
Unconjugated
ELISA (1:8000) Western Blot (0.5-1.5 ug/mL) The optimal working dilution should be determined by the end user.
CARD17
Liquid
CARD17
A synthetic peptide corresponding to C-terminus of human CARD17.
100 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only