Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC50, Goat, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of CCDC50.

Supplier:  Abnova Corporation PAB6706

Catalog No. 89-132-617


Add to Cart

Description

Description

This gene encodes a soluble, cytoplasmic, tyrosine-phosphorylated protein with multiple ubiquitin-interacting domains. Mutations in this gene cause nonsyndromic, postlingual, progressive sensorineural DFNA44 hearing loss. In mouse, the protein is expressed in the inner ear during development and postnatal maturation and associates with microtubule-based structures. This protein may also function as a negative regulator of NFkb signaling and as an effector of epidermal growth factor (EGF)-mediated cell signaling. Alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq

Sequence: CFSKSESSHKGFHYK
Specifications

Specifications

CCDC50
Polyclonal
Unconjugated
ELISA (1:32000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5-10 ug/mL) The optimal working dilution should be determined by the end user.
CCDC50
C3orf6/DFNA44/YMER
Goat
Antigen affinity purification
RUO
152137
Human
Liquid
ELISA, Immunohistochemistry (PFA fixed)
0.5 mg/mL
Goat polyclonal antibody raised against synthetic peptide of CCDC50.
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
NP_848018.1;NP_777568.1
CCDC50
A synthetic peptide corresponding to human CCDC50.
100 μg
Primary
This antibody is expected to recognize both reported isoforms of human Ymer protein.
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only