Learn More
CDON, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of CDON.
Supplier: Abnova Corporation PAB17230
Description
CDON and BOC (MIM 608708) are cell surface receptors of the immunoglobulin (Ig)/fibronectin type III (FNIII; see MIM 135600) repeat family involved in myogenic differentiation. CDON and BOC are coexpressed during development, form complexes with each other in a cis fashion, and are related to each other in their ectodomains, but each has a unique long cytoplasmic tail.[supplied by OMIM
Sequence: C-KNRQQNTIQKYDPSpecifications
CDON | |
Polyclonal | |
Unconjugated | |
ELISA (1:32000) The optimal working dilution should be determined by the end user. | |
CDON | |
CDO/MGC111524/ORCAM | |
Goat | |
Antigen affinity purification | |
RUO | |
50937 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of CDON. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
NP_058648.4 | |
CDON | |
A synthetic peptide corresponding to amino acids at internal region of human CDON. | |
100 μg | |
Primary | |
Human | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only