Learn More
Goat anti-Choline Oxidase, Biotin, Polyclonal, Novus Biologicals™
Goat Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP1777050.1MG
Catalog No. NBP17770501
Add to cart
Description
Description
Choline Oxidase Polyclonal antibody specifically detects Choline Oxidase in Bacteria samples. It is validated for Western Blot, ELISA, Immunoprecipitation.
Specifications
Specifications
Choline Oxidase
Biotin
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide
Choline oxydase
Goat
IgG
0.1 mg
Store at 4C. Do not freeze.
Polyclonal
Bacteria
ELISA, Immunoprecipitation, Western Blot
Western Blot 1:500 - 1:5000, ELISA 1:5000 - 1:20000, Immunoprecipitation 1:100
Purified
CHOX_ALCSP
Choline Oxidase Native Protein (Alcaligenes gunus).
Delipidation and Defibrination
RUO
Primary
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Goat Serum as well as purified and partially purified Choline Oxidase (Alcaligenes species). Cross reactivity against Choline Oxidase from other sources is unknown.