Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHRNB3, Goat, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of CHRNB3.

Supplier:  Abnova Corporation PAB7469

Catalog No. 89-133-280


Add to Cart

Description

Description

Sequence: C-SRHVKKEHFISQ
Specifications

Specifications

CHRNB3
Polyclonal
Unconjugated
ELISA (1:16000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user.
CHRNB3
CHRNB3
A synthetic peptide corresponding to human CHRNB3.
100 μg
Primary
Human
Liquid
ELISA, Western Blot
0.5 mg/mL
Goat polyclonal antibody raised against synthetic peptide of CHRNB3.
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
NP_000740.1
Goat
Antigen affinity purification
RUO
1142
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only