Learn More
Goat anti-Collagen Type I, Biotin, Polyclonal, Abnova™
Manufacturer:  Abnova Corporation PAB14217
Catalog No. 89-137-856
Description
Description
Specifications
Specifications
Collagen Type I
Biotin
Conjugated
Human type I collagen.
Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
Polyclonal
ELISA, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot
In PBS, pH 7.4 (0.09% sodium azide)
Goat
200μg
Primary
Human