Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL13, Goat, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of CXCL13.

Manufacturer:  Abnova Corporation PAB18603

Catalog No. 89-120-161


Add to cart

Description

Description

B lymphocyte chemoattractant, independently cloned and named Angie, is a CXC chemokine strongly expressed in the follicles of the spleen, lymph nodes, and Peyer's patches. It preferentially promotes the migration of B lymphocytes (compared to T cells and macrophages), apparently by stimulating calcium influx into, and chemotaxis of, cells expressing Burkitt's lymphoma receptor 1 (BLR-1). It may therefore function in the homing of B lymphocytes to follicles. [provided by RefSeq

Sequence: KNKSIVCVDPQAE
Specifications

Specifications

CXCL13
0.5 mg/mL
Goat polyclonal antibody raised against synthetic peptide of CXCL13.
In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA)
NP_006410.1
ANGIE|ANGIE2|BCA-1|BCA1|BLC|BLR1L|SCYB13
Goat
Antigen affinity purification
RUO
Primary
10563
ELISA
Unconjugated
ELISA (1:32000) The optimal working dilution should be determined by the end user.
CXCL13
Liquid
CXCL13
A synthetic peptide corresponding to amino acids at internal region of human CXCL13.
100 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only