Learn More
CYP450 2B4, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against CYP450 2B4.
Manufacturer:  Abnova Corporation PAB13659
Catalog No. 89-137-650
Add to cart
Description
Description
Specifications
Specifications
CYP450 2B4
Unconjugated
Western Blot (1:100) The optimal working dilution should be determined by the end user.
Liquid
Rabbit CYP450 2B4.
RUO
Primary
This goat polyclonal antibody specifically reacts with rabbit CYP450 2B4. Reacts as a single band with liver microsomes prepared from rabbits that were induced to express multiple forms of P450's.
ELISA, Western Blot
Goat polyclonal antibody raised against CYP450 2B4.
In buffer containing 0.05% sodium azide
Goat
1 mL
Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Rabbit, Rat