Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAG1, Goat, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of DAG1.

Manufacturer:  Abnova Corporation PAB18550

Catalog No. 89-120-109


Add to cart

Description

Description

Dystroglycan is a laminin binding component of the dystrophin-glycoprotein complex which provides a linkage between the subsarcolemmal cytoskeleton and the extracellular matrix. Dystroglycan 1 is a candidate gene for the site of the mutation in autosomal recessive muscular dystrophies. The dramatic reduction of dystroglycan 1 in Duchenne muscular dystrophy leads to a loss of linkage between the sarcolemma and extracellular matrix, rendering muscle fibers more susceptible to necrosis. Dystroglycan also functions as dual receptor for agrin and laminin-2 in the Schwann cell membrane. The muscle and nonmuscle isoforms of dystroglycan differ by carbohydrate moieties but not protein sequence. Alternative splicing results in multiple transcript variants

Sequence: C-HVGKHEYFMHATDK
Specifications

Specifications

DAG1
Polyclonal
Unconjugated
ELISA (1:128000) Western Blot (0.2-0.6 ug/mL) The optimal working dilution should be determined by the end user.
DAG1
156DAG/A3a/AGRNR/DAG
Goat
Antigen affinity purification
RUO
1605
Human
Liquid
ELISA, Western Blot
0.5 mg/mL
Goat polyclonal antibody raised against synthetic peptide of DAG1.
In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA)
NP_004384.3
DAG1
A synthetic peptide corresponding to amino acids at internal region of human DAG1.
100 μg
Primary
Reported variants represent identical protein: NP_001159400.1, NP_004384.3.
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only