Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DARC, Goat, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of DARC.

Supplier:  Abnova Corporation PAB6873

Catalog No. 89-132-756


Add to Cart

Description

Description

The protein encoded by this gene is a glycosylated membrane protein and a nonspecific receptor for several chemokines. The encoded protein is the receptor for the human malarial parasites Plasmodium vivax and Plasmodium knowlesi. Polymorphisms in this gene are the basis of the Duffy blood group system. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: HRAELSPSTENSSQLDFED-C
Specifications

Specifications

DARC
Polyclonal
Unconjugated
ELISA (1:16000) Western Blot (0.1-0.3 ug/mL) Immunohistochemistry (3 ug/mL) The optimal working dilution should be determined by the end user.
DARC
CCBP1/CD234/Dfy/FY/GPD/GpFy/WBCQ1
Goat
Antigen affinity purification
RUO
2532
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
ELISA, Flow Cytometry, Immunohistochemistry (PFA fixed), Western Blot
0.5 mg/mL
Goat polyclonal antibody raised against synthetic peptide of DARC.
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
NP_002027.2
DARC
A synthetic peptide corresponding to human DARC.
100 μg
Primary
Human
Liquid
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only