Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDAH1, Goat, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of DDAH1.

Supplier:  Abnova Corporation PAB6093

Catalog No. 89-132-137


Add to Cart

Description

Description

This gene belongs to the dimethylarginine dimethylaminohydrolase (DDAH) gene family. The encoded enzyme plays a role in nitric oxide generation by regulating cellular concentrations of methylarginines, which in turn inhibit nitric oxide synthase activity. [provided by RefSeq

Sequence: TCCSVLINKKVDS
Specifications

Specifications

DDAH1
Polyclonal
Unconjugated
ELISA (1:16000) Western Blot (0.1-0.3 ug/mL) The optimal working dilution should be determined by the end user.
DDAH1
DDAH/FLJ21264/FLJ25539
Goat
Antigen affinity purification
RUO
23576
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
ELISA, Western Blot
0.5 mg/mL
Goat polyclonal antibody raised against synthetic peptide of DDAH1.
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
NP_0.36269.1
DDAH1
A synthetic peptide corresponding to human DDAH1.
100 μg
Primary
Human
Liquid
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only