Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DIAPH3, Goat, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of DIAPH3.

Supplier:  Abnova Corporation PAB7395

Catalog No. 89-133-213

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

Sequence: C-RTPMPKDVRQS
Specifications

Specifications

DIAPH3
Polyclonal
Unconjugated
ELISA (1:64000) The optimal working dilution should be determined by the end user.
DIAPH3
DKFZp434C0931/DKFZp686A13178/DRF3/Dia2/FLJ34705/diap3
Goat
Antigen affinity purification
RUO
81624
Human
Liquid
ELISA
0.5 mg/mL
Goat polyclonal antibody raised against synthetic peptide of DIAPH3.
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
NP_0010.35982.1;NP_112194.2
DIAPH3
A synthetic peptide corresponding to human DIAPH3.
100 μg
Primary
This antibody is expected to recognize both reported isoforms (NP_0010.35982.1 and NP_112194.2).
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only