Learn More
diphtheria toxoid, Goat, FITC, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against diphtheria toxoid.
Manufacturer:  Abnova Corporation PAB14229
Catalog No. 89-137-864
Add to cart
Description
Description
Specifications
Specifications
diphtheria toxoid
FITC
The optimal working dilution should be determined by the end user.
Liquid
Purified diphtheria toxoid.
RUO
Primary
Demonstrated reactivity to purified diphtheria toxin/toxoid.
ELISA, Gel Shift
Goat polyclonal antibody raised against diphtheria toxoid.
In 10mM PBS, pH 7.2 (10mg/mL BSA, 0.09% sodium azide)
Goat
1 mL
Store in the dark at 4°C. For long term storage store at -20°C.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Bacteria