Learn More
IgG (H+L) Goat anti-Donkey, Biotin, Invitrogen™
Goat Polyclonal Secondary Antibody
Manufacturer:  Invitrogen PA128737
Catalog No. PIPA128737
Add to cart
Description
Description
PA1-28737 detects Donkey IgG from donkey samples. PA1-28737 has been successfully used in ELISA, immunohistochemistry (frozen tissue), immunohistochemistry (paraffin tissue), Western blot applications. The PA1-28737 immunogen is: Full length native donkey IgG (purified).
Thermo Scientific Anti-Equine secondary antibodies are affinity-purified antibodies with well-characterized specificity for equine (horse) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Specifications
Donkey IgG (H+L)
Polyclonal
Biotin
Liquid
Full length native donkey IgG (purified).
IgG fraction
RUO
Secondary Antibody
Polyclonal
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunomicroscopy, Western Blot
2 mg/mL
0.02M potassium phosphate with 0.15M NaCl, 10mg/mL BSA and 0.01% sodium azide; pH 7.2
Goat
IgG
1 mg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Secondary
Donkey