Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Goat anti-Donkey IgG (H+L) Secondary Antibody, FITC, Invitrogen™
Goat Polyclonal Secondary Antibody
Supplier: Thermo Scientific PA128734
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Donkey IgG (H+L) Polyclonal antibody specifically detects Donkey IgG (H+L) in Donkey samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunomicroscopy.Specifications
Donkey IgG (H+L) | |
Polyclonal | |
FITC | |
Goat | |
Antigen affinity chromatography | |
RUO | |
Donkey | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunomicroscopy | |
2 mg/mL | |
0.02M potassium phosphate with 0.15M NaCl, 10mg/mL BSA and 0.01% sodium azide; pH 7.2 | |
Full length native Donkey IgG (purified). | |
1 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction